Dry sponge
Leverancier: 3M Medical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MMMEBP133ESEA 274 EUR
MMMEBP133ES
Dry sponge
Schoonmaakdoekjes Sponzen
This dry sampling sponge is made of biocide-free cellulose to maintain organism viability. The cellulose material is effective for capturing particulates in dry sampling scenarios.
  • Can be used with any number of in-house or pre-mixed buffers and sampling solutions
  • Suitable for use in dry-only facilities where dry sampling is required
  • Sample bag has a writable surface to note sample details and for quick, easy identification
  • Simplified packaging convenience minimizes handling and preparation and reduces waste
Leveringsinformatie: Supplied with 18 oz (532 ml) printed sample bag.