ORF Clones, GABA B receptor 1 (GABBR1), human tagged, TrueORF®
Leverancier: OriGene
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ORIGRC214114EA 1690 EUR
ORIGRC214114
ORF Clones, GABA B receptor 1 (GABBR1), human tagged, TrueORF®
Vectoren, Plasmiden en Bibliotheken
GABA B receptor 1 (GABBR1), human tagged ORF clone was engineered to express the complete ORF with an expression tag. Expression varies depending on the nature of the gene.
This gene encodes a receptor for gamma-aminobutyric acid (GABA), which is the main inhibitory neurotransmitter in the mammalian central nervous system. This receptor functions as a heterodimer with GABA(B) receptor. Defects in this gene may underlie brain disorders such as schizophrenia and epilepsy. Alternative splicing generates multiple transcript variants, but the full-length nature of some of these variants has not been determined.
The ORF clone is ion-exchange column purified, transfection-ready dried plasmid DNA.
Leveringsinformatie: Includes 2 vector sequencing primers.