ORF Clones, GABA B receptor 2 (GABBR2), human tagged, TrueORF Gold
Leverancier: OriGene
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ORIGRC219404EA 1690 EUR
ORIGRC219404
ORF Clones, GABA B receptor 2 (GABBR2), human tagged, TrueORF Gold
Vectoren, Plasmiden en Bibliotheken
GABA B receptor 2 (GABBR2), human tagged ORF clone was engineered to express the complete ORF with an expression tag. Expression varies depending on the nature of the gene.
The ORF clone is ion-exchange column purified, transfection-ready dried plasmid DNA.
he multi-pass membrane protein encoded by this gene belongs to the G-protein coupled receptor 3 family and GABA-B receptor subfamily. The GABA-B receptors inhibit neuronal activity through G protein-coupled second-messenger systems, which regulate the release of neurotransmitters, and the activity of ion channels and adenylyl cyclase. This receptor subunit forms an active heterodimeric complex with GABA-B receptor subunit 1, neither of which is effective on its own. Allelic variants of this gene have been associated with nicotine dependence.
Leveringsinformatie: Include 2 vector sequencing primers.