Bar magnet
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
761-0051EA 29 EUR
BREU51050
Bar magnet
This bar magnet is made of alnico alloy and has different colored poles.
  • Dimensions: 60×13×5 mm