Blood diluting pipettes, acc. to Malassez-Potain
Leverancier: Karl Hecht
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HECH40359702EA 6.5 EUR
HECH40359702 HECH40359701
Blood diluting pipettes, acc. to Malassez-Potain
Blood diluting pipettes for red corpuscles (erythrocytes) or white corpuscles (leucocytes).
  • Fused blue graduation “ASSISTENT-PERMANENT”
  • Division at 1, 2, 3, 4 and 5