Autoclavable cleanroom goggles, Superblast Autoclave
Leverancier: Bollé Safety
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BOLESUPBLCLAVEEA 25.23 EUR
BOLESUPBLCLAVE
Autoclavable cleanroom goggles, Superblast Autoclave
Ruimzichtbrillen
These autoclavable cleanroom goggles offer a panoramic vision and an unequalled comfort, with PC lens and PLATINUM® coating included.
  • Ventilated frame and high-comfort
  • Overflow chute for liquids
  • Adjustable strap and clean
  • Fits over prescription glasses
  • Compatible with half-mask respirator
PLATINUM® coated PC lens: Coating on both sides ensures high scratch-resistance (1.4 cd/m²), high resistance to the most aggressive chemicals and slows the appearance of fogging.
Weight: 92 g