SpCas9 nuclease
Leverancier: Polyplus-transfection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
722-100EA 0 EUR
722-100
SpCas9 nuclease
Transfectiereagentia
SpCas9 nuclease sequence is carefully optimized to improve its nuclear import, nuclease editing efficiency, and to reduce off-targeting effects
  • High genome editing efficiency with jetCRISPR™
  • Especially designed for transfection
  • Minimize off target-effects using RNP approach
  • Better Cas9 activity control by using RNP
Choosing the optimal Cas9 sequence is key in enhancing targeted nuclear delivery of the Cas9 protein with the gRNA. Polyplus-transfection SpCas9 nuclease leads to higher genome editing then other commercially available Cas9 proteins using jetCRISPR™ transfection reagent.
SpCas9 nuclease is developed to reach the highest genome editing efficiency with our RNP transfection reagent jetCRIPSR™.
Concentration 10 µg/µL
Certificaten: Since 2002, the company is ISO 9001 certified and since then this certification has been successfully renewed every three years.
Bestelinformatie: 1.5 ml of jetCRISPR™ transfection reagent is sufficient to perform ~300 transfections in 6-well plates. Contact your local Avantor sales office for more details.
Opgelet: Storage Temperature - 20 °C