Netherlands
Order Entry
Filler Vials and Vial Module for use with CoolCell LX and CoolCell FTS30 Containers, Corning® CoolCell®
Filler Vials and Vial Module for use with CoolCell LX and CoolCell FTS30 Containers, Corning® CoolCell®
Supplier:  Corning
   Sustainable Options Available
Certificates
undefined
Filler Vials and Vial Module for use with CoolCell LX and CoolCell FTS30 Containers, Corning® CoolCell®
Supplier:  Corning
   Sustainable Options Available
Certificates
Specifications
  • Catalog No:
  • 479-1961
  • 479-1852
  • 479-1962
  • Environmentally Preferable:
  • Safer Human and Environmental Health
Specifications
Related Information
Product Information
  • Catalog #
    Supplier #
    For
    Pk
    Availability
    Price
  • CoolCell® 2 mL filler vials
    CoolCell® 2 mL filler vials
    479-1961
    432076
    Use with CoolCell LX and CoolCell FTS30 containers
    6
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 28,25 €
    28,25 €
    1 * 6 ST
       28,25 €
    Supplier Article Number: 432076
    Local Article Number: CORN432076
    Specifications:
    Specifications
    Environmentally Preferable
  • Removable cryogenic vial module, holds 30 cryogenic vials (1.0 mL or 2.0 mL)
    Removable cryogenic vial module, holds 30 cryogenic vials (1.0 mL or 2.0 mL)
    479-1852
    432078
    CoolCell FTS30
    10
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 116,13 €
    116,13 €
    1 * 10 ST
       116,13 €
    Supplier Article Number: 432078
    Local Article Number: CORN432078
    Specifications:
      Safer Human and Environmental Health
    More
    Specifications
    Environmentally Preferable
    Safer Human and Environmental Health
      Safer Human and Environmental Health
  • CoolCell® 5 mL filler vials
    CoolCell® 5 mL filler vials
    479-1962
    432077
    Use with CoolCell 5mL LX container
    6
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 28,25 €
    28,25 €
    1 * 6 ST
       28,25 €
    Supplier Article Number: 432077
    Local Article Number: CORN432077
    Specifications:
    Specifications
    Environmentally Preferable