Chamber furnaces, forced convection
Leverancier: Nabertherm
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
NABENA-0361D2EA 6330 EUR
NABENA-0361D2 NABENA--5040F2 NABENA--5041D2 NABENA--0641C2 NABENA-2561D2 NABENA--0341D1 NABENA-1261D2 NABENA-0661D2 NABENA--0340F1 NABENA--2540F2 NABENA--1241D2 NABENA--0161C1 NABENA--2541D2 NABENA-5061D2 NABENA--1240F2 NABENA-0361C2 NABENA--0640F2 NABENA--0641D2 NABENA-2561C2 NABENA-1261C2 NABENA--0341C1 NABENA--2541C2 NABENA--5041C2 NABENA-0661C2 NABENA--1241C2 NABENA-5061C2 NABENA--0161B1
Chamber furnaces, forced convection
Ovens Ovens met hoge temperatuur
The outstanding temperature uniformity of these chamber furnaces with air circulation provides ideal process conditions for annealing, curing, solution annealing, artificial ageing, preheating, or soft annealing and brazing.
  • Stainless steel air-baffles in the furnace for optimum air circulation
  • Swing door hinged on the right hand side
  • Horizontal air circulation
  • Optimum air distribution enabled by high flow speeds
  • NTLog Basic for Nabertherm controller: Recording of process data with USB-flash drive
  • Temperature uniformity up to ±4 °C according to DIN 17052-1 (±7 °C for N 15/65 HA models)
These forced convection chamber furnaces are equipped with a suitable annealing box for soft annealing of copper or tempering of titanium, and also for annealing steel under non-flammable protective or reaction gases.
The B400 standard controller is equipped with LC display, featuring 5 freely adjustable programs, 4 segments and 2 extra functions. The optional C440 controller has 10 programs with 20 segments each. Only the NA 15/65 bench-top model has the B410 or C450 controller, these controllers are PID microprocessor controlled with a self-diagnostic system and integrated process documentation.
Bestelinformatie: Supplied with base frame, frame sheet and rails for two additional trays, (except N 15/65 HA models without frame sheet). Optional accessories such as pneumatic lift door, controlled cooling options, manual in- and outlet exhaust system, motor driven inlet and outlet flaps. Please contact your Avantor sales office for more information on options.
Opgelet: Model NA 15/65 HA is a bench-top unit; all other models are floor standing models.