Wire baskets
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
41178.KKEA 82.58 EUR
41178.KK 41177.KK 41178.KD
Wire baskets
Manden Draadmanden
Made from Tungsten
  • Wire Baskets for laboratory needs for holding high temperature samples.
For vacuum deposition