pH/conductivity electrode, Eutech
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
EUTEECCOMBI03MEA 286 EUR
EUTEECCOMBI03M
pH/conductivity electrode, Eutech
Elektroden Multiparameter elektroden
Combined pH electrode and 2-pin stainless steel conductivity electrode with 15 cm ABS guard. For use with CyberScan PC 10.