D-(+)-Galactose
Leverancier: MP Biomedicals
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ICNA114021002EA
0
EUR
ICNA114021002
D-(+)-Galactose
D-(+)-Galactose
Formule:
C₆H₁₂O₆ MW: 180,16 g/mol Kookpunt: 527 °C (1013 hPa) Smeltpunt: 163…169 °C Dichtheid: 1,5 g/cm³ (20 °C) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00151230 CAS nummer: 59-23-4 EINECS: 200-416-4 Merck Index: 13,04356 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...