Portable gas leak detector, MiniTemp MT4
Leverancier: PerkinElmer
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PERCN9306074EA 194 EUR
PERCN9306074
Portable gas leak detector, MiniTemp MT4
The MiniTemp MT4 is capable of displaying thermal measurement readings using easy point and shot infrared technology in a pocket size configuration.
  • Light weight and portable
  • Displays thermal measurement readings in °C or °F
  • Typical distance to target (spot) up to 1.5 m (4 ft)
Great for instrument thermal test confirmation, including GC injector port and detector measurements, thermostatted LC vials, and enzymatic hydrolysis baths.
Height: 152,4 mm
Leveringsinformatie: Supplied with single dot laser sighting to assist with aiming and 9 V battery.
Opgelet: Recalibration is not available.