(±)-1-Octen-3-ol for synthesis, Sigma-Aldrich®
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
M-Clarity™ quality level = MQ200
8.18543.9999
(±)-1-Octen-3-ol for synthesis, Sigma-Aldrich®
(±)-1-Octen-3-ol
Formule:
C₈H₁₆O MW: 128,21 g/mol Kookpunt: 174 °C (1013 hPa) Smeltpunt: –49 °C Dichtheid: 0,83 g/cm³ (20 °C) Vlampunt: 68 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00004589 CAS nummer: 3391-86-4 EINECS: 222-226-0 UN: 2810 ADR: 6.1,II |
Specification Test Results
Solubility | (20 °C) slightly soluble |
Melting Point | -49 °C |
Molar Mass | 128.21 g/mol |
Boiling Point | 173 - 175 °C (1013 hPa) |
Vapor Pressure | 1 hPa (20 °C) |
Flash Point | 68 °C |
Refractive Index | 1.4378 (20 °C, 589 nm) |
Explosion Limit | 0.9 - 8 %(V) |
Density | 0.83 - 0.85 g/cm3 (20 °C) |
pH | (H2O) neutral |
Ignition Point | 245 °C |
Viscosity Kinematic | 5.3 mm2/s (23 °C) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...