D-(-)-Aspartic acid
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BACE4030335.0100EA
0
EUR
BACE4030335.0100
BACE4030335.0025
D-(-)-Aspartic acid
D-(-)-Aspartic acid
Formule:
C₄H₇NO₄ MW: 133,1 g/mol Smeltpunt: 251 °C |
MDL Nummer:
MFCD00063081 CAS nummer: 1783-96-6 EINECS: 217-234-6 Merck Index: 13,00845 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...