(S)-(+)-Hydratropic acid 99%
Leverancier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
ACRO264652500EA
46.3
EUR
ACRO264652500
ACRO264650010
(S)-(+)-Hydratropic acid 99%
(S)-(+)-Hydratropic acid
Formule:
C₉H₁₀O₂ MW: 150,18 g/mol Kookpunt: 115 °C (1 mmHg) Smeltpunt: 29…32 °C Dichtheid: 1,1 g/cm³ Vlampunt: >110 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00063139 CAS nummer: 7782-24-3 |
Specification Test Results
Appearance | Off-white Low melting solid |
Infrared spectrum | Conforms |
Melting point | 29 to 32 °C |
GC | ≥98.5 % |
Specific optical rotation | +74° to +76° (20 °C, 589 nm) (c=3, CHCl3) |