Dimethyl malonate
Leverancier: EHRENSTORFER GMBH
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
EHERC14731500EA
56.6
EUR
EHERC14731500
Dimethyl malonate
Dimethyl malonate
Formule:
C₅H₈O₄ MW: 132,12 g/mol Kookpunt: 181 °C (1013 hPa) Smeltpunt: –62 °C Dichtheid: 1,15 g/cm³ (20 °C) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00008460 CAS nummer: 108-59-8 EINECS: 203-597-8 Merck Index: 12,06169 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...