Netherlands
Order Entry
Succinic acid for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
undefined
Succinic acid for synthesis, Sigma-Aldrich®
Supplier:  Merck
Specification Test Results
  • Solubility:
    58 g/l (20 °C)
  • Melting Point:
    185 - 190 °C
  • Molar Mass:
    118.09 g/mol
  • Bulk Density:
    940 kg/m3
  • Boiling Point:
    235 °C (1013 hPa)
  • Flash Point:
    206 °C
  • Density:
    1.56 g/cm3 (20 °C)
  • pH:
    2.7 (10 g/l, H2O, 20 °C)
  • Ignition Point:
    470 °C
Specification Test Results
  •  
    Pack type
    Pk
  • 8.22260.0250
    8.22260.0250
    Plastic bottle
    250 g
    1 * 250 g
    1 * 250 g - 38,30 €
    38,30 €
    1 * 250 g
       38,30 €
  • 8.22260.1000
    8.22260.1000
    Plastic bottle
    1 kg
    1 * 1 kg
    1 * 1 kg - 77,00 €
    77,00 €
    1 * 1 kg
       77,00 €
  • 8.22260.9050
    8.22260.9050
    50 kg
    1 * 50 kg
    1 * 50 kg