ASTM precision thermometers
Leverancier: Amarell
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AMARA300788-FLEA 69.5 EUR
AMARA300788-FL AMARA300010-FL AMARA300790-FL AMARA300280-FL AMARA300150-FL AMARA301160-FL AMARA301250-FL AMARA301150-FL AMARA301020-FL AMARA300260-FL AMARA300798-FL AMARA300780-FL AMARA300220-FL AMARA301280-FL AMARA300800-FL AMARA301140-FL AMARA301870-FL AMARA301220-FL AMARA301348-FL AMARA301014-FL AMARA300770-FL AMARA300210-FL AMARA300080-FL AMARA301210-FL AMARA300020-FL AMARA301860-FL AMARA300290-FL AMARA301350-FL AMARA300160-FL AMARA301170-FL AMARA301130-FL AMARA300070-FL AMARA301260-FL
ASTM precision thermometers
Thermometers Analoge thermometers
These models are suitable for government testing or official calibration.
  • Solid stem thermometer
  • White backed
Certificaten: Compliant with ASTM.