Phenylacetic acid ≥99%
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
A14881.36EA
29.8
EUR
A14881.36
Phenylacetic acid ≥99%
Phenylacetic acid
Formule:
C₈H₈O₂ MW: 136,15 g/mol Kookpunt: 265 °C Smeltpunt: 76…79 °C Dichtheid: 1,100 Vlampunt: 132 °C (270 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00004313 CAS nummer: 103-82-2 EINECS: 203-148-6 Merck Index: 12,07422 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...