Netherlands
Order Entry
Diethyl fumarate
Irritant
Diethyl fumarate
Catalog # EHERC12606600
Supplier:  EHRENSTORFER
CAS Number:  
Irritant
Diethyl fumarate
Catalog # EHERC12606600
Supplier:  EHRENSTORFER
CAS Number:  
Supplier Article Number: C12606600
Local Article Number: EHERC12606600
Price
71,40 €
Log in for availability
Unit of Measure 1 * 0,25 g

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
 :172.18 g/mol
Boiling Pt:219 °C (1013 hPa)
Melting Pt:1…2 °C
Density:1,05 g/cm³ (20 °C)
Flash Pt:91 °C
Storage Temperature:Ambient
EINECS:210-819-7
 :GHS07
CAS Number:623-91-6
Formula:C₈H₁₂O₄
MDL Number:MFCD00064455