5-Chloro-2-methoxybenzoic acid
Leverancier: Thermo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MAYBSB01454.1EA 0 EUR
MAYBSB01454.1
5-Chloro-2-methoxybenzoic acid
5-Chloro-2-methoxybenzoic acid
Formule: C₈H₇ClO₃
MW: 186,59 g/mol
MDL Nummer: MFCD00017546
CAS nummer: 3438-16-2
EINECS: 222-343-7