3,5-Dichlorosalicylic acid ≥98%
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
B23641.06EA
17.4
EUR
B23641.06
3,5-Dichlorosalicylic acid ≥98%
3,5-Dichlorosalicylic acid
Formule:
C₇H₄Cl₂O₃ MW: 207,01 g/mol Kookpunt: 319 °C (1013 hPa) Smeltpunt: 222…225 °C |
MDL Nummer:
MFCD00002442 CAS nummer: 320-72-9 EINECS: 206-281-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...