Houd mijn sessie geopend
Eindigt in 
De sessie is verlopen
Uw sessie is verlopen. Voor uw veiligheid bent u uitgelogd.
Wilt u opnieuw inloggen?

3-(Methylthio)phenyl isothiocyanate ≥97%

Leverancier: Alfa Aesar

Purity: 97%
CAS No.: 51333-80-3
Molecular Formula: C8H7NS2
Formula Weight: 181.28
Flash Point: 110°(230°F)
Refractive Index: 1.6910
Boiling Point: 144-145°/8mm
Beilstein Reference: 2208144
UN No.: 2922
MDL No.: MFCD00041079


Formule: C₈H₇NS₂
MW: 181,28 g/mol
Kookpunt: 130 °C (1013 hPa)
Dichtheid: 1,22 g/cm³ (20 °C)
MDL Nummer: MFCD00041079
CAS nummer: 51333-80-3


 

Order

Expand All / Collapse All
VPE VWR catalogus nummer Hoeveelheid
1 g L12999.03
5 g L12999.06
Expand All / Collapse All
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Specificaties

Formulegewicht 181.28
CAS No. 51333-80-3
Zuiverheid 97%
Flash point >110°(230°F)
Kookpunt 144-145°/8mm
Molecular formula C8H7NS2
MDL No. MFCD00041079
UN No. 2922

WRITE A REVIEW

Need help?

  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.

VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.

Please confirm the industry you work in

Example: Pharmaceutical

Please confirm your role

Example: Clinical Trial Assistant

Please confirm your location

Example: New York, NY

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'X' if unknown for VWR Chemicals. Enter a space if unknown for Merck Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Of
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Omschrijving Catalogus nummer Availability Unit Uw prijs Price Per Hoeveelheid