4-Aminofenol ≥99.0% for synthesis, Sigma-Aldrich®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-Clarity™ quality level = MQ200
8.00421.9050EA 0 EUR
8.00421.9050 8.00421.0250 8.00421.9999 8.00421.9025 8.00421.0005 8.00421.9100 8.00421.1000
4-Aminofenol ≥99.0% for synthesis, Sigma-Aldrich®
4-Aminofenol
Formule: H₂NC₆H₄OH
MW: 109,13 g/mol
Kookpunt: 284 °C (1013 hPa) (ontbinding)
Smeltpunt: 190 °C
Vlampunt: 195 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00007869
CAS nummer: 123-30-8
EINECS: 204-616-2
ADR: 6.1,III
Merck Index: 13,00461
https://nl.vwr.com/stibo/web/std.lang.all/04/69/2300469.jpg
Order Now
Specification Test Results
Assay (HClO₄) ≥ 99.0 %
Identity (IR) passes test