Houd mijn sessie geopend
Eindigt in 
De sessie is verlopen
Uw sessie is verlopen. Voor uw veiligheid bent u uitgelogd.
Wilt u opnieuw inloggen?

2-Aminothiophenol for synthesis

Leverancier: Merck
Waarschuwing


Formule: C₆H₇NS
MW: 125,19 g/mol
Kookpunt: 234 °C (1013 hPa)
Smeltpunt: 26 °C
Dichtheid: 1,17 g/cm³ (20 °C)
Brandingspunt: 79 °C
Storage Temperature: Koelkast
MDL Nummer: MFCD00007702
CAS nummer: 137-07-5
EINECS: 205-277-3
UN: 1759
ADR: 8,II
Specification Test Results
Solubility 0.027 g/l
Melting Point 26 °C
Molar Mass 125.18 g/mol
Boiling Point 234 °C (1013 hPa)
Vapor Pressure 1 hPa (69 °C)
Flash Point 79 °C
Refractive Index 1.6405 (20 °C, 589 nm)
Density 1.17 g/cm3 (20 °C)

login om uw prijzen en beschikbaarheid te zien
  Onthoud mij op dit apparaat
 

Order

Expand All / Collapse All
VPE VWR catalogus nummer Hoeveelheid
100 ml 8.01335.0100
25 ml 8.01335.0025
Expand All / Collapse All
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

WRITE A REVIEW

Need help?

  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.

VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.

Please confirm the industry you work in

Example: Pharmaceutical

Please confirm your role

Example: Clinical Trial Assistant

Please confirm your location

Example: New York, NY

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'X' if unknown for VWR Chemicals. Enter a space if unknown for Merck Chemicals

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Of
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Omschrijving Catalogus nummer Availability Unit Uw prijs Price Per Hoeveelheid