Density gradient medium, OptiPrep™
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
D1556-250MLEA 593 EUR
D1556-250ML
Density gradient medium, OptiPrep™
Media voor celscheiding
Density gradient medium is suitable for cell and subcellular organelle isolation, virus purification, and isolation of lipoproteins and macromolecules.
OptiPrep™ Density Gradient Medium has been used in the non enveloped virion isolation from bronchoalveolar lavage.