VWR® Vial Kit for Karl Fischer Titration
Leverancier: VWR
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VWR®
548-2398EA 368 EUR
548-2398
VWR® Vial Kit for Karl Fischer Titration
flesjes
This vial kit is compatible with oven sample processors 774.
  • Tight and reliable vial/cap combination
  • Keep the water content constant, as well for longer transport/storage times