Transduction reagent, Viro-MICST™
Supplier: OZ BIOSCIENCES
Certificates
About this item
Viro-MICST™ transduction reagent allows genetically modifying cells directly on magnetic cell purification columns. Based on i-MICST™ Technology (Integrated Magnetic Immuno-Cell Sorting and Transfection/Transduction), this technology combines cell isolation and genetic modification in one simple and reliable integrated system. For efficient and selective gene delivery to target cells, Viro-MICST™ can be used in combination with any viral vector and allows you to reduce cell manipulation steps and save time and material.
- Highly efficient
- Rapid, simple, and ready to use
- Suitable for all cells
Store at +4 °C.
Delivery information: Shipped at room temperature.
Specifications
- Cell specific:CD4+, CD8+, T lymphocytes
Specifications
Related Information
Product Information
- Catalogus #Leverancier #PkAvailabilityPrice
- Viro-MICST™ transduction reagentSupplier Article Number: VMX250Local Article Number: OZBIVMX250
- Viro-MICST™ transduction reagentSupplier Article Number: VMX500Local Article Number: OZBIVMX500
- Viro-MICST™ transduction reagentViro-MICST™ transduction reagentOZBIVMX1000VMX10001.000 µlLog in for availability1 * 1.000 µl841,36 €Supplier Article Number: VMX1000Local Article Number: OZBIVMX1000
Frequently Bought Together