Karl Fischer reagents, buffer for bases, HYDRANAL™
Leverancier: Honeywell Research Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HYDRANAL™
37859-6X500MLEA 244.06 EUR
37859-6X500ML 37859-500ML 37859-1L 37859-6X1L
Karl Fischer reagents, buffer for bases, HYDRANAL™
Karl Fischer titranten en reagentia
Liquid buffer medium for Karl Fischer titration in alkaline samples (based on salicylic acid).