Netherlands
Order Entry
Two-way adaptors, PEEK™
Two-way adaptors, PEEK™
  OMNI002306
 :  OMNIFIT
undefined
Two-way adaptors, PEEK™
  OMNI002306
 :  OMNIFIT
 : 002306
 : OMNI002306
39,00 €
  1 * 1 ST
  • Thread:
    1/4 28 F, 10 - 32 M
  • Description:
    Threaded Adaptors, Natural, PEEK
  • Seq:
    01
  • Pk:
    1
Omnifit® threaded adaptors are a useful range of problem solving parts that enable to convert thread sizes quickly and easily, accepting a ¹/₄"-28 UNF male fitting at one end and a range of male and female thread types at the other.