Labels for TLS 2200®/TLS PC Link™ printers, type B-499
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BRDY011211EA 101.9 EUR
BRDY011211 BRDY062266 BRDY018509 BRDY062252 BRDY062273 BRDY018503 BRDY018504 BRDY018502
Labels for TLS 2200®/TLS PC Link™ printers, type B-499
Etiketten
Nylon cloth labels for the TLS2200® and TLS PC Link™ thermal transfer portable printers.
  • Matt finish
  • For smooth, textured/rough or highly textured/low surface energy surfaces
TLS2200® parts can also be printed on the BMP™71 label printer provided the roll adapter (M71-ADAPT) is used.