Alcometer Professional fuel cell breathalyser
Leverancier: SureScreen Diagnostics
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SUREALCOMPROEA 440 EUR
SUREALCOMPRO
Alcometer Professional fuel cell breathalyser
The Alcometer Professional is a fuel cell breathalyser offering reliable, accurate results using the gold standard in breath alcohol testing. It is perfect for mass screening and workforce testing.
  • Long life
  • Increased accuracy with an extra digit for reading results
  • Screen mode which allows rapid screening every 5 to 10 seconds
  • Flow sensor to indicate donor is using the device correctly
  • Backlit displays for use at night
  • Increased testing speed
  • Warning lights for positive results
  • 6 months/500 uses between calibrations
Two models available: Results in BrAC or results in promilles.
Leveringsinformatie: Kit comes with 3 mouthpieces, battery and a heavy duty case.