ProClean extra, J.T.Baker®
Leverancier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BAKR3862.1000EA 0 EUR
BAKR3862.1000
ProClean extra, J.T.Baker®
Verpakking: Verpakt in poly (5 l) en polycube (20 l).