Lanyard, DBI-SALA® EZ-Stop™ Expander™, 3M™
Leverancier: 3M
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CPSD1245534EA 301.3 EUR
CPSD1245534 CPSD1245541
Lanyard, DBI-SALA® EZ-Stop™ Expander™, 3M™
Lanyard with elasticated webbing reduces length by up to 500 mm when not under tension.
  • Reduced trip or snarling hazard
  • Leg material 35 mm polyester webbing
Certificaten: EN 355