Hoist system and Masts, DBI-SALA®
Leverancier: DBI/Sala
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CPSD8568040EA 0 EUR
CPSD8568040 CPSD8568509 CPSD8568006 CPSD8568003 CPSD8568004 CPSD8568387 CPSD8568001 CPSD8568386 CPSD8568002 CPSD8568385 CPSD8568384 CPSD8568000 CPSD8568383 CPSD8568382
Hoist system and Masts, DBI-SALA®
Hoist systems are designed for manhole and confined space entry/retrieval applications.
  • Constructed of lightweight materials including high-strength aluminium
  • Davit pivots for ease-of-rescue and adjustment for overhead clearance restrictions
  • Lower base adjusts to fit most standard entries
Many other portable and fixed bases are available to suit any jobsite need.
Weight capacity: 205 kg
Certificaten: EN 795 Class-B
Bestelinformatie: Winch and SRL ordered separately.
Leveringsinformatie: Hoist system includes short/ long offset adjustable offset upper mast, 838 mm lightweight lower mast and 3-piece, lightweight base and winch mounting bracket.