Wound closure system, Autoclip®
Leverancier: Harvard Apparatus
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Autoclip®
HARV34-0554EA 0 EUR
HARV34-0554 HARV34-0557 HARV34-0556 HARV34-0555
Wound closure system, Autoclip®
Eerste-hulpbenodigdheden Wound Clips
This Autoclip® wound closure system provides a rapid, easy to use method to securely close wounds. When the wound is healed, simply remove the clip using the handy removal forcep.
  • Provides maximum holding force with minimal trauma
Automatic, spring-fed applicators permit rapid, unassisted application.
Leveringsinformatie: Starter set includes 100 wound clips, applicator, and removal forcep.