Silane adhesive slides, KF Frost
Leverancier: Klinipath
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
631-1238EA 314 EUR
631-1238
Silane adhesive slides, KF Frost
Object- en dekglaasjes Adhesie-objectglaasjes
These silane adhesive slides are specially designed for thermal transfer printers.
Certificaten: CE certified according to IVDR (2017/746).