Netherlands
Order Entry
Ethylene oxide
Ethylene oxide
 :  Restek
undefined
Ethylene oxide
 :  Restek
  • CAS No.:
    75-21-8
  • Formula:
    C₂H₄O
  • Description:
    Ethylene oxide
  •  
    Concentration
    Pk Info
    Pk
  • Ethylene oxide
    Ethylene oxide
    REST36005
    36005
    500 μg/ml in dimethyl sulphoxide
    1 ml/ampoule
    1
    1 * 1 ST
    1 * 1 ST - 58,00 €
    58,00 €
    1 * 1 ST
       58,00 €
     : 36005
     : REST36005
  • Ethylene oxide
    Ethylene oxide
    REST30620
    30620
    50 mg/ml in methylene chloride
    1 ml/ampoule
    1 mL
    1 * 1 mL
    1 * 1 mL - 131,00 €
    131,00 €
    1 * 1 mL
       131,00 €
     : 30620
     : REST30620
This product has been blocked by your organization. Please contact your purchasing department for more information