BTEX standard (6 components)
Supplier: Restek
Certificates
About this item
Contains: Benzene (71-43-2); Ethylbenzene (100-41-4); Toluene (108-88-3); m-Xylene (108-38-3); o-Xylene (95-47-6); p-Xylene (106-42-3)
Product Information
- Catalog #Supplier #Pk InfoPkAvailabilityPrice
- BTEX standard with 200 μg/ml each component in P and T methanolBTEX standard with 200 μg/ml each component in P and T methanolREST30051300511 ml/ampule1Log in for availability1 * 1 ST58,00 €Supplier Article Number: 30051Local Article Number: REST30051
- BTEX standard with 2000 μg/ml each component in P and T methanol (m-xylene and p-xylene at 1000 μg/ml)BTEX standard with 2000 μg/ml each component in P and T methanol (m-xylene and p-xylene at 1000 μg/ml)REST30488304881 ml/ampule1Log in for availability1 * 1 ST71,50 €Supplier Article Number: 30488Local Article Number: REST30488
- BTEX standard with 2000 μg/ml each component in P and T methanolBTEX standard with 2000 μg/ml each component in P and T methanolREST30213302131 ml/ampule1 mLLog in for availability1 * 1 mL71,50 €Supplier Article Number: 30213Local Article Number: REST30213