Barcode reader, wireless, VisionMate™
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VisionMate®
479-2237EA 3257 EUR
479-2237
Barcode reader, wireless, VisionMate™
Barcodesystemen
The VisionMate™ wireless barcode reader is a versatile sample tracking instrument that decodes 2D and linear barcodes and QR codes. The wireless reader utilises Bluetooth™ technology to allow mobility in the lab, cold room or in the field to yield efficient sample identification and tracking, and for direct input into a paired workstation, LIMS or portable device.
  • For use with 24-, 48-, 96-, 384- tube racks
  • Mobility while transmitting barcode data
  • Versatile format compatibility
  • Compatible with all linear (1D), 2D Matrix and Nunc tube and rack formats
  • Direct data output to many applications using keyboard wedge
  • Ambient light diffuser
  • Clear high-contrast screen
The VisionMate™ wireless reader is capable of reading any barcode of ANSI Grade D or better including Thermo Scientific™ linear and 2D barcodes and those of most major manufacturers, making it well suited for multi-facility applications.
Leveringsinformatie: Includes wireless barcode reader with CodeXML software, USB cable and Belkin Bluetooth device with associated software, USB wall adapter with US/EU/UK/AU compatible plugs.