TBX (Tryptone Bile X‑glucuronide) agar, Chromocult®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1.16122.0500EA 369 EUR
1.16122.0500
TBX (Tryptone Bile X‑glucuronide) agar, Chromocult®
Media voor microbiologie Gedehydrateerde media voor microbiologie
For the enumeration, detection and identification of β-glucuronidase positive Escherichia coli from food and feeding stuff as well as from environmental samples in the area of food production and food handling.
Certificaten: Complies with the specifications given by ISO 16649 part 1 and part 2 and with those given by EN ISO 16649-3.