Benchtop autoclaves, Elara Class B
Leverancier: Tuttnauer
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
TUTTELARA11-DEA 6190 EUR
TUTTELARA11-D
Benchtop autoclaves, Elara Class B
These models are reliable and robust Class B autoclaves designed for the needs of modern clinics where autoclaves are expected to meet sophisticated international standards and operate non-stop every day. Suitable for the sterilisation processing needs of medical, dental and veterinary clinics.
  • Self-locking door designed to easily close/open door with one hand
  • Bright LCD panel for easy reading, with multilingual interface (26 languages)
  • Automatic safety shutoff prevents overheating
  • Maintenance free with reduced running cost
  • Built-in system for checking water quality
Durable 316L type stainless steel chamber and electro-polish finish door.
Class B performance is achieved with a built-in vacuum pump used for fractionated pre-vacuum air removal eliminating air pockets from
all load types and maximising efficient steam penetration throughout the entire load. After the sterilisation stage the vacuum pump is used for post-vacuum drying.
Features a user-friendly three button keypad enables the user to easily browse through the sterilisation cycle program settings and user directories.
Certificaten: Complies with ISO 9001:2008, EN ISO 13485, AAMI/ANSI ST-55:2010,EN 13060, EN 61010-1, EN 61326-1, EN 61010-2-40, EN 17665-1.