Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
Login
/
Register
Netherlands
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
0
0,00 €
Order Entry
Home
Enzymes
Exonuclease I
Exonuclease I
Exonuclease I
:
Thermo Fisher Scientific
Exonuclease I
:
Thermo Fisher Scientific
Enzyme name:
Exonuclease I
Concentration:
20 U/µl
Pk Info
Pk
EN0581
EN0581
4000 units
1 KIT
1 * 1 Kit
1 * 1 Kit - 110,00 €
110,00 €
-
+
1 * 1 Kit
110,00 €
-
+
: EN0581
: FERMEN0581
EN0582
EN0582
20000 units
1 KIT
1 * 1 Kit
1 * 1 Kit - 443,00 €
443,00 €
-
+
1 * 1 Kit
443,00 €
-
+
: EN0582
: FERMEN0582