Netherlands
Order Entry
Serum vials, borosilicate glass
Serum vials, borosilicate glass
Supplier:  DWK Life Sciences
Certificates
undefined
Serum vials, borosilicate glass
Supplier:  DWK Life Sciences
Certificates
Product Information
  • Catalog #
    Supplier #
    Capacity
    Ø×H
    Colour
    Cap included
    Cap Size
    Availability
    Price
  • Vials with A/S finish
    Vials with A/S finish
    SCERSP62113D-2
    SP62113D-2
    2,00 ml
    15×40 mm
    Clear
    No
    13 mm
    Log in for availability
    Log in for availability
    1 * 288 ST
    1 * 288 ST - 1 340,00 €
    1 340,00 €
    1 * 288 ST
       1 340,00 €
    Supplier Article Number: SP62113D-2
    Local Article Number: SCERSP62113D-2
    Specifications:
    Specifications
    Environmentally Preferable
  • Vials with A/S finish
    Vials with A/S finish
    SCERSP62113D-312
    SP62113D-312
    3,50 ml
    18×45 mm
    Clear
    No
    13 mm
    Log in for availability
    Log in for availability
    1 * 288 ST
    1 * 288 ST - 48,10 €
    48,10 €
    1 * 288 ST
       48,10 €
    Supplier Article Number: SP62113D-312
    Local Article Number: SCERSP62113D-312
    Specifications:
    Specifications
    Environmentally Preferable
  • Vials with A/S finish
    Vials with A/S finish
    SCERSP62121D-5
    SP62121D-5
    5,00 ml
    23×47 mm
    Clear
    No
    20 mm
    Log in for availability
    Log in for availability
    1 * 864 ST
    1 * 864 ST - 765,00 €
    765,00 €
    1 * 864 ST
       765,00 €
    Supplier Article Number: SP62121D-5
    Local Article Number: SCERSP62121D-5
    Specifications:
    Specifications
    Environmentally Preferable
  • Vials with A/S finish
    Vials with A/S finish
    SCERSP62121D-10
    SP62121D-10
    10,00 ml
    25×54 mm
    Clear
    No
    20 mm
    Log in for availability
    Log in for availability
    1 * 864 ST
    1 * 864 ST - 1 210,00 €
    1 210,00 €
    1 * 864 ST
       1 210,00 €
    Supplier Article Number: SP62121D-10
    Local Article Number: SCERSP62121D-10
    Specifications:
    Specifications
    Environmentally Preferable
  • Vials with A/S finish
    Vials with A/S finish
    SCERSP62121U-10
    SP62121U-10
    10,00 ml
    24×50 mm
    Clear
    No
    20 mm
    Log in for availability
    Log in for availability
    1 * 1.085 ST
    1 * 1.085 ST - 683,00 €
    683,00 €
    1 * 1.085 ST
       683,00 €
    Supplier Article Number: SP62121U-10
    Local Article Number: SCERSP62121U-10
    Specifications:
    Specifications
    Environmentally Preferable
  • Vials with A/S finish
    Vials with A/S finish
    SCER6212D-20
    6212D-20
    20,00 ml
    30×57 mm
    Clear
    No
    20 mm
    Log in for availability
    Log in for availability
    1 * 144 ST
    1 * 144 ST - 237,00 €
    237,00 €
    1 * 144 ST
       237,00 €
    Supplier Article Number: 6212D-20
    Local Article Number: SCER6212D-20
    Specifications:
    Specifications
    Environmentally Preferable
  • Vials with A/S finish
    Vials with A/S finish
    WHEA4042467
    4042467
    10,00 ml
    − mm
    Clear
    No
    20 mm
    Log in for availability
    Log in for availability
    1 * 2.100 ST
    1 * 2.100 ST - 3 110,00 €
    3 110,00 €
    1 * 2.100 ST
       3 110,00 €
    Supplier Article Number: 4042467
    Local Article Number: WHEA4042467
    Specifications:
    Specifications
    Environmentally Preferable