Hygenic mask, silv-Air 8213
Leverancier: Uvex
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
UVEX8708.213EA 42.1 EUR
UVEX8708.213
Hygenic mask, silv-Air 8213
These FFP 2 folding masks with exhalation valve, featuring an innovative design and an optimised shape.
  • Hydrophobic material coating
  • Seamless headband for a comfortable and secure fit
  • Adjustable nose piece and sealing lip around the nose and chin ensures a comfortable and secure positioning of the mask