Hygenic masks, silv-Air 8313
Leverancier: Uvex
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
UVEX8708.313EA 87.8 EUR
UVEX8708.313
Hygenic masks, silv-Air 8313
FFP 3 folding mask with exhalation valve features an innovative design and an optimised shape. The adjustable nose piece and all-round ultrafine porous sealing lip ensures a comfortable and secure positioning of the masks.
  • Hydrophobic material coating
  • Four-point headband attachment for a comfortable, individual and secure fit
  • Exhalation valve to further reduce breathing resistance by helping to minimise moisture build up inside the mask