Netherlands
Order Entry
Decon 90 laboratory detergent
Decon 90 laboratory detergent
Catalog # PERC09923462
Supplier:  PerkinElmer
Decon 90 laboratory detergent
Catalog # PERC09923462
Supplier:  PerkinElmer
Supplier Article Number: 09923462
Local Article Number: PERC09923462
Price
69,50 €
Log in for availability
Unit of Measure 1 * 1 L
Specifications
  • Description:
    Decon 90
  • Pk:
    1 L
Specifications
About this item
Surface active cleaning agent/radioactive decontaminant for laboratory, medical and industrial applications.
  • Milky white, non-viscous liquid, having faint odour
  • A liquid concentrate, for dilution with water
  • Phosphate-free, bactericidal, non-flammable and totally rinsable
  • Problem contaminants can be treated by increasing the solution strength
  • For critical applications, use demineralised water throughout
Used for cleaning and/or decontaminating glassware, ceramics, rubbers, most plastics, stainless steel and ferrous metals.
Cleaning time can be reduced by heating the solution (40 to 50 °C) or using in an ultrasonic cleaning bath. The cleaned items must be rinsed thoroughly as soon as they are removed from the cleaning solution, and dried. Three agitated rinses are recommended in water to ensure removal of all traces of contamination and cleaning agent.
Certifications: OECD 301E (ISO method 7827 - 1986 (E)) and satisfies the criteria in BS EN 1040 1997 for bacterial activity.
Packaging: Supplied in high density polyethylene container, with tamper-evident screw caps.
Caution: NOT suitable for domestic applications. Being alkaline, DECON 90 is NOT suitable for use on non-ferrous metals, notably aluminium and zinc, or on polycarbonate.