About this item
HiTrap MabSelect SuRe PCC chromatography columns are prepacked with MabSelect SuRe™ pcc protein A resin for screening and small-scale mAb purification in continuous bioprocessing using periodic counter-current chromatography (PCC).
- Convenient prepacked column for fast, reproducible, and easy small-scale purifications
- High binding capacity, ~ 60 mg human IgG/ml resin at 2,4 min residence time
- High productivity through capture of large mass of monoclonal antibodies in a short period of time
- Highly concentrated elution pools for operating flexibility and small unit operations
- Allows cost-effective cleaning with 0,1 to 0,5 M NaOH over hundreds of purification cycles
- High protein ligand stability reduces ligand leakage
HiTrap MabSelect SuRe PCC packed columns (1 and 5 ml) are suited for small scale antibody purifications, as well as optimization of methods and parameters, such as sample load and protein binding conditions. These protein A columns provide fast, reproducible, and easy separations in a convenient format. The columns are optimally utilized when they are connected to liquid chromatography systems such as the AKTA systems.
The alkali-stabilised, protein A-derived ligand gives the resin greater stability than conventional protein A-based resins under the alkaline conditions used in cleaning-in-place (CIP) protocols. Consequently, the possibility of cleaning with cost-effective reagents such as sodium hydroxide (NaOH) will improve process economy.
In addition, the use of NaOH for cleaning efficiently inactivates bacteria, mold, and yeast, with improved product quality as a result.
Specifications
- Catalog No:
- 17-5491-12
- 17-5491-11
- Max. pressure:
- 5 bar
- 5 bar
- Storage:
- 2...8 °C, 20% Ethanol
- 2...8 °C, 20% Ethanol
Specifications
Related Information
Cytiva CyDye Licensing Statements Cytiva General Licensing Statements
Additional Product Details
Product Information
- Catalog #Supplier #Column hardwareBed dimensionsCapacityFlow rateFlow velocityAvailabilityPrice
- HiTrap™ MabSelect SuRe™ pccHiTrap™ MabSelect SuRe™ pcc17-5491-1117-5491-11PP7×25 mm1 ml<4 ml/min≤600 cm/h1 * 5 ST987,00 €: 17-5491-11: GEHE17-5491-11
Specifications
Environmentally PreferableMax. pressure5 barStorage2...8 °C, 20% Ethanol - HiTrap™ MabSelect SuRe™ pccHiTrap™ MabSelect SuRe™ pcc17-5491-1217-5491-12PP16×25 mm5 ml<20 ml/min≤600 cm/h1 * 1 ST922,00 €: 17-5491-12: GEHE17-5491-12
Specifications
Environmentally PreferableMax. pressure5 barStorage2...8 °C, 20% Ethanol