Chemical resistant gloves, Tychem® NT450
Leverancier: DuPont
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Tychem®
112-5482EA 0 EUR
112-5482 112-5483 112-5485
Chemical resistant gloves, Tychem® NT450
Handschoenen
These cotton/polyester gloves with double nitrile coating protect the hand and forearm from oils, hydrocarbons, grease and abrasion.
  • Cotton liner absorbs perspiration for comfortable extended wear, with good mechanical resistance
  • Impermeable for working in damp or greasy environments
  • Rough textured finish over surface of hand
  • Ergonomic fit
  • Natural rubber free
  • Wrist protection
Ideal for heavy duty work or longer exposure to hazardous chemicals and solvents.
Certificaten: EN 388:2016 (4112X), EN ISO 374-1:2016 Type A (JKLOPT), EN ISO 374-5 (VIRUS), Cat. III
Food contact approved.